Skip to Content
Merck
All Photos(2)

Documents

SAB1301227

Sigma-Aldrich

ANTI-HAS2 (CENTER) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

HAS2, Hyaluronan synthase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

conjugate

unconjugated

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

63566 Da

species reactivity

human

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:50-1:100
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HAS2(3037)

Related Categories

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chi Feng et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 77(8), 1582-1593 (2019-03-25)
Idiopathic condylar resorption (ICR) is an aggressive form of osteoarthritis that is frequently observed in adolescent female patients. We hypothesized that an estrogen-mediated pathway may contribute to ICR development. An enzyme-linked immunosorbent assay was used to detect the levels of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service