Skip to Content
Merck
All Photos(2)

Key Documents

SAB1300959

Sigma-Aldrich

Anti-ALOX5AP (center) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-ALOX5AP, Anti-Arachidonate 5-lipoxygenase-activating protein, Anti-FLAP, Anti-MK-886-binding protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$458.00

$458.00


Estimated to ship on23 May 2025



Select a Size

Change View
100 μG
$458.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$458.00


Estimated to ship on23 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALOX5AP(241)

General description

ALOX5AP is a protein which, with 5-lipoxygenase, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.

Immunogen

ALOX5AP (NP_001620, 91-127)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the Center region of human ALOX5AP.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service