SAB1300760
Anti-PARP3 (C-term) antibody produced in rabbit
saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
Synonym(s):
Anti-Adprtl3, Anti-NAD+ ADP-ribosyltransferase 3 PARP-3, Anti-Parp3, Anti-Poly (ADP-ribose polymerase)-like 3, full insert sequence), NOD-derived CD11c +ve dendritic cells cDNA, RIKEN full-length enriched library, clone:F630107G12 product:ADP-ribosyltransferase (NAD+
Select a Size
$404.00
Estimated to ship on01 June 2025
Select a Size
About This Item
$404.00
Estimated to ship on01 June 2025
biological source
rabbit
conjugate
unconjugated
antibody form
saturated ammonium sulfate (SAS) precipitated
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human, mouse
technique(s)
indirect ELISA: 1:1000
western blot: 1:100-1:500
NCBI accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PARP3(235587)
General description
The previously assigned protein identifier Q8CFB8 has beenmerged into Q3ULW8. Full details can be found on the UniProt database.
Immunogen
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of mouse PARP3.
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service