Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300696

Sigma-Aldrich

Anti-OIT3(L) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-LZP, Anti-Liver-specific zona pellucida domain-containing protein, Anti-OIT3, Anti-Oncoprotein-induced transcript 3 protein [Precursor]

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$404.00

$404.00


Check Cart for Availability


Select a Size

Change View
100 μG
$404.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$404.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OIT3(170392)

General description

Liver-specific ZP domain-containing protein (OIT3) is a liver-specific protein involved possibly in hepatocellular function and development. Oncoprotein induced transcript 3 (OIT3) is also known as liver-specific zona pellucida domain-containing protein and is composed of 545 amino acid residues. It is expressed in the liver and secreted into the blood. OIT3 possesses a D8C (domain with conserved eight cysteines) domain, an amino-terminal signal peptide and three epidermal growth factor (EGF)-like domains. The cDNA of the protein consists of 2,255bp. The gene encoding OIT3 is localized on human chromosome 10q21.3, spans 40kb and has nine exons.

Immunogen

OIT3 (NP_689848, )
This antibody is generated from rabbits immunized with a GST tag human OIT3 recombinant protein.

Biochem/physiol Actions

Oncoprotein induced transcript 3 (OIT3) may have a role in liver development and function. It has been shown to be downregulated in hepatocellular carcinoma.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service