Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300394

Sigma-Aldrich

Anti-MARK4 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-KIAA1860, Anti-MAP/microtubule affinity-regulating kinase 4, Anti-MAP/microtubule affinity-regulating kinase-like 1, Anti-MARKL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MARK4(57787)

General description

MARK4 contains an N-terminal serine/threonine kinase domain, a central ubiquitin-associated domain, and a C-terminal KA1-associated kinase domain. RT-PCR analysis detects upregulated expression of the gene for MARK4 in nearly all clinical hepatocellular carcinoma cells. Northern blot analysis reveals ubiquitous expression of a 3.6-kb transcript, with highest expression in testis. Immunofluorescence microscopy demonstrates homogeneous cytoplasmic expression. Colony-forming assays show that MARK4 antisense reduces the growth of SNU475 cells. It has been suggested that MARK4 provides a growth advantage to cells.

Immunogen

MARK4 (NP_11360, 15-45)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human MARK4.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivier E Pardo et al.
EMBO reports, 17(4), 570-584 (2016-02-18)
Here, we show that miR-515-5p inhibits cancer cell migration and metastasis. RNA-seq analyses of both oestrogen receptor receptor-positive and receptor-negative breast cancer cells overexpressing miR-515-5p reveal down-regulation of NRAS, FZD4, CDC42BPA, PIK3C2B and MARK4 mRNAs. We demonstrate that miR-515-5p inhibits

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service