Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300329

Sigma-Aldrich

Anti-MLL3 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-HALR, Anti-Histone-lysine N-methyltransferase, H3 lysine-4 specific MLL3, Anti-Homologous to ALR protein, Anti-MLL3, Anti-Myeloid/lymphoid or mixed-lineage leukemia protein 3 homolog

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$404.00

$404.00


Check Cart for Availability


Select a Size

Change View
100 μG
$404.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$404.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MLL3(58508)

General description

The SET domain is a conserved C-terminal domain that characterizes proteins of the MLL family, including MLL3. The MLL SET domain is a histone H3 Lys4 (K4)-specific methyltransferase whose activity is stimulated with acetylated H3 peptides. MLL3 maps to 7q36, a chromosome region frequently deleted in myeloid leukaemia. The deduced MLL3 4,911-amino acid protein is more closely related to MLL2 than to MLL1 or MLL4. MLL3 has 6 plant homeodomain (PHD) fingers preceded by a cys-rich ZNF1 domain in its N terminus; a high mobility group (HMG) box, an ATPase alpha-beta signature, and a leucine zipper motif in its central region; and 2 C-terminal FY (phe-tyr) motifs and the SET domain preceded by a ZNF2 domain in its C terminus. The predicted protein also contains several putative nuclear localization motifs. Isoform II lacks ZNF1 and the PHD.

Immunogen

MLL3 (NP_733751, 316-350)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human MLL3.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service