Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300315

Sigma-Aldrich

Anti-ABCB5 (N-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-ABCB5, Anti-ABCB5 P-gp, Anti-ATP-binding cassette sub-family B member 5, Anti-P-glycoprotein ABCB5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$404.00

$404.00


Check Cart for Availability


Select a Size

Change View
100 μG
$404.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$404.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCB5(340273)

General description

ABCB5 (ATP binding cassette subfamily B member 5) gene is located in human chromosome 7p21.1. The encoded protein belongs to the subfamily B multidrug resistance and ATP-binding cassette superfamily of active transporters. The encoded protein have two identical halves, each contains a hydrophobic region with six transmembrane segments and a nucleotide-binding region. Expression of this gene is observed in prostate and testis and in human melanoma tumor-initiating cells.
ABCB5 belongs to the ATP-binding cassette (ABC) transporter superfamily of integral membrane proteins. These proteins participate in ATP-dependent transmembrane transport of structurally diverse molecules ranging from small ions, sugars, and peptides to more complex organic molecules.

Immunogen

ABCB5 (NP_848654, 3-35)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human ABCB5.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Flow cytometry/Cell sorting (1 paper)

Biochem/physiol Actions

ABCB5 (ATP binding cassette subfamily B member 5) is a cell surface marker for melanoma-initiating cells, on cellular metabolism. ABCB5 significantly regulates glycolysis, glutaminolysis and phospholipid metabolism. It induces stem cell differentiation, stem cell immortality and anti-apoptotic activity. The gene is upregulated in many cancer subtypes, including breast cancer. Recent studies reveal that ABCB5 is a novel marker for limbal stem cell and is needed for corneal development and repair. ABCB5 shows resistance against taxanes and anthracyclines, the anti-cancer drugs.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juntao Yao et al.
Oncology research, 25(3), 305-316 (2017-03-11)
ABCB5 belongs to the ATP-binding cassette (ABC) superfamily, which is recognized for playing a role in the failure of chemotherapy. ABCB5 has also been found to be overexpressed at the transcriptional level in a number of cancer subtypes, including breast
Norbert W Lutz et al.
PloS one, 11(8), e0161803-e0161803 (2016-08-26)
We present a pilot study aimed at determining the effects of expression of ATP-binding cassette member B5 (ABCB5), a previously described marker for melanoma-initiating cells, on cellular metabolism. Metabolic profiles for two groups of human G3361 melanoma cells were compared
Shingo Kondo et al.
BMC pharmacology & toxicology, 16, 37-37 (2015-12-17)
Previously, we have demonstrated that human ABCB5 is a full-sized ATP-binding cassette transporter that shares strong homology with ABCB1/P-glycoprotein. ABCB5-transfected cells showed resistance to taxanes and anthracyclines. Herein, we further screened ABCB5 substrates, and explored the mechanism of resistance. Sensitivity
Vladan Milosevic et al.
International journal of cancer, 146(1), 192-207 (2019-05-21)
Malignant pleural mesothelioma (MPM) is a tumor with high chemoresistance and poor prognosis. MPM-initiating cells (ICs) are known to be drug resistant, but it is unknown if and how stemness-related pathways determine chemoresistance. Moreover, there are no predictive markers of
Passara Jongkhajornpong et al.
Scientific reports, 6, 20541-20541 (2016-02-05)
ATP-binding cassette subfamily B member 5 (ABCB5) is a new member of the ATP-binding cassette superfamily and has been reported as a novel marker for limbal stem cell (LSC), which is essential for corneal homeostasis. ABCB5 expression has also been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service