Skip to Content
Merck
All Photos(1)

Key Documents

SAB1105806

Sigma-Aldrich

Anti-ASB13 (215-225) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Ankyrin repeat and SOCS box-containing 13

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$389.00

$389.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$389.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$389.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen predicted mol wt ~30 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ASB13(79754)

General description

Ankyrin repeat and SOCS box containing 13 (ASB13) is part of the ankyrin repeat and SOCS box (ASB) family and belongs to the suppressor of cytokine signaling (SOCS) box protein superfamily. It might interact with proteins like the RNA polymerase III subunit A and single-stranded DNA-binding protein. The gene encoding ASB13 is localized on human chromosome 10p15.1.

Immunogen

synthetic peptide corresponding to amino acids 215-225 of human ASB13

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ASB proteins interact with Cullin5 and Rbx2 to form E3 ubiquitin ligase complexes.
Kohroki J, et al.
FEBS Journal, 579(30), 6796-6802 (2005)
Genome-wide Cardiovascular Disease Risk of Sudden Deaths Between 5-40 Years in Turkish Population Investigation of Factors.
Cavlak G O C O, et al.
The Journal of Experimental Medicine, 6(12), 45-47 (2016)
Christina Aaen Andresen et al.
The Journal of biological chemistry, 289(4), 2043-2054 (2013-12-18)
The ankyrin and SOCS (suppressor of cytokine signaling) box (ASB) family of proteins function as the substrate recognition subunit in a subset of Elongin-Cullin-SOCS (ECS) E3 ubiquitin ligases. Despite counting 18 members in humans, the identity of the physiological targets

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service