Skip to Content
Merck
All Photos(1)

Key Documents

SAB1103260

Sigma-Aldrich

Anti-DCAF15 (567-580) antibody produced in rabbit

Anti-DCAF15 (567-580) antibody produced in rabbit
1 of 1 reviewers received a sample product or took part in a promotion

IgG fraction of antiserum

Synonym(s):

Anti-C19orf72, Anti-DDB1 and CUL4 associated factor 15

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$565.00

$565.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$565.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$565.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~66 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DCAF15(90379)

General description

DCAF15 (DNA damage binding protein1 (DDB1) and cullin4 (CUL4) associated factor 15) gene in human chromosome, is mapped to 19p13.12. DCAF15 lacks protein interaction domain.
DCAF15 protein is a substrate receptor for E3 ligase. This protein is a part of the cullin 4-RING (CRL4)-DCAF15 E3 ubiquitin ligase.

Immunogen

synthetic peptide corresponding to amino acids 567-580 of human DCAF15

Application

Anti-DCAF15 (567-580) antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

DCAF15 (DNA damage binding protein 1 (DDB1) and cullin4 (CUL4) associated factor 15) is the vertebrate ortholog of Saccharomyces pombe chromosome licensing and DNA replication factor 2 (Cdt2) and is required for destruction of Cdt1 during S phase and after DNA damage in human cells.
DCAF15 protein plays a role in RNA splicing and cell apoptosis of various cancer cell lines. This protein is also involved in ubiquitination and degradation of splicing factor RNA binding motif protein 39 (RBM39).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A family of diverse Cul4-Ddb1-interacting proteins includes Cdt2, which is required for S phase destruction of the replication factor Cdt1
Jin J, et al.
Molecular Cell, 23(5), 709-721 (2006)
Expanding the spectrum of rearrangements involving chromosome 19: A mild phenotype associated with a 19p13.12-p13. 13 deletion
Marangi G, et al.
American Journal of Medical Genetics. Part A, 158(4), 888-893 (2012)
Xiao Dong et al.
Aging, 13(7), 10603-10618 (2021-04-10)
Epithelial-mesenchymal transition (EMT) is an evolutionarily conserved developmental program that has been implicated in tumorigenesis and confers metastatic properties upon cancer cells. ZEB1 is a master transcription factor that activates the EMT process in various cancers. ZEB1 is reportedly degraded
Tabitha C Ting et al.
Cell reports, 29(6), 1499-1510 (2019-11-07)
Indisulam and related sulfonamides recruit the splicing factor RBM39 to the CRL4-DCAF15 E3 ubiquitin ligase, resulting in RBM39 ubiquitination and degradation. Here, we used a combination of domain mapping and random mutagenesis to identify domains or residues that are necessary

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

    • Review 1
    • Vote 1
    1 out of 5 stars.

    Rubbish - does not work

    unspecific bands on WB - extremely weak band at 72kDa but no change upon knockdown with siRNA

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for taking the time to leave a review! We are sorry to hear that your experience did meet expectations. We would like to learn more about your experience to see if there is anything we can do to help correct this issue. When you have a moment, please contact us by visiting https://www.sigmaaldrich.com/support/customer-support and submit a Report Product Issues ticket. We look forward to hearing from you soon!

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service