Skip to Content
Merck
All Photos(2)

Key Documents

S9875

Sigma-Aldrich

Sodium choleate

Synonym(s):

Choleic Acid Sodium Salt, Desoxycholic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161900
NACRES:
NA.25
Pricing and availability is not currently available.

description

anionic

technique(s)

electrophoresis: suitable

Application

Sodium choleate has been used in a study to assess the resistance of esophageal adenocarcinoma to bile salt. [1] It has also been used in a study to investigate the role of a putative tyrosine-O-sulfate receptor in the targeting and/or intracellular transport of tyrosine-sulfated proteins. [2]

Other Notes

This is a loosely used term applied to a crude ox bile extract which contains the sodium salts of taurocholic, glycocholic, deoxycholic, and cholic acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Resistance of Esophageal Adenocarcinoma to Bile Salt Insult is Associated with Manganese Superoxide Dismutase Expression
Schiffman, S., et al.
The Journal of Surgical Research, 171, 8-8 (2011)
M C Liu et al.
Cytotechnology, 23(1-3), 143-149 (1997-01-01)
By employing the affinity gel fraction technique, we have detected a 175 kDa tyrosine-O-sulfate (TyrS)-binding protein in sodium choleate extracts of the microsomal membrane fractions of bovine liver and pancreas, as well as canine liver and pancreas. Western blot analysis
Suguru Yamasaki et al.
Scientific reports, 9(1), 177-177 (2019-01-19)
During infection, Salmonella senses and responds to harsh environments within the host. Persistence in a bile-rich environment is important for Salmonella to infect the small intestine or gallbladder and the multidrug efflux system AcrAB-TolC is required for bile resistance. The
Giovanni Sorrentino et al.
Gastroenterology, 159(3), 956-968 (2020-06-03)
Renewal and patterning of the intestinal epithelium is coordinated by intestinal stem cells (ISCs); dietary and metabolic factors provide signals to the niche that control ISC activity. Bile acids (BAs), metabolites in the gut, signal nutrient availability by activating the

Questions

  1. I would like to know that is this product suitable for neonatal mouse experiment?

    1 answer
    1. This product has not been tested in neonatal mouse studies. The researcher would have to establish suitability. There is a cell culture tested product option, C9282, that may be a better option. The product is not sterile, but has been tested for cytotoxicity in mammalian cell lines. See below for a link to this product.
      https://www.sigmaaldrich.com/product/sigma/c9282

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service