Skip to Content
Merck
All Photos(1)

Documents

S8321

Sigma-Aldrich

Anti-Sonic Hedgehog antibody, Mouse monoclonal

clone SH154, purified from hybridoma cell culture

Synonym(s):

Anti-Shh

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SH154, monoclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

concentration

~2 mg/mL

technique(s)

immunohistochemistry: 1:500 using formaldehyde-fixed human tissue sections
indirect ELISA: 2 μg/mL
western blot: 2 μg/mL using recombinant mouse SHH peptide

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SHH(6469)
mouse ... Shh(20423)
rat ... Shh(29499)

General description

The Hedgehog (Hh) family of secreted proteins consists of sonic hedgehog (Shh), desert hedgehog (Dhh) and Indian hedgehog (Ihh). The Hh signaling regulates cell migration and tissue construction during embryogenesis and reconstruction in adults. Shh protein is secreted by Purkinje cells in the cerebellum with both autocrine and paracrine functions. The Shh signaling is mediated by the interaction of Shh with transmembrane proteins, Patched and Smoothened. The Shh pathway cross-talks Notch and Wnt pathways and acts via downstream targets such as cyclins, γ catenin, myc and other transcription factors. The Hh family members have multiple roles as morphogens, cell survival factors, setting anterior-posterior axis and digit formation during development of limb and axon guidance factors during the development of brain and nervous system. Hedgehog signaling has been implicated in childhood tumors of pancreas, liver, thyroid, gastrointestinal tract, neuroblastomas and medulloblastomas
Anti-Sonic Hedgehog antibody recognizes human, rat and mouse Shh.

Specificity

Mouse monoclonal clone SH154 anti-Sonic Hedgehog antibody recognizes human, rat, and mouse Shh.

Application

Anti-Sonic Hedgehog antibody may be used for immunoblotting at a working concentration of ~2 μg/ml. A working antibody concentration of ~2 μg/ml is recommended for ELISA. For immunohistochemistry a dilution of 1:500 may be used.
Mouse monoclonal clone SH154 anti-Sonic Hedgehog antibody may be used in ELISA, immunoblotting, and immunohistochemistry. It is used as a probe to determine the presence and roles of Sonic Hedgehog in processes such as patterning during embryogenesis.

Biochem/physiol Actions

Sonic Hedgehog (Shh) is an important cell signaling molecule expressed during embryonic development. Shh is involved in patterning of the developing embryonic systems such as the nervous system, somites, and limbs. The N-terminal peptide of Shh is released by autoproteolysis and functions through interactions with a multicomponent receptor complex containing the transmembrane proteins Patched and Smoothened. Shh protein is expressed in key embryonic tissues such as the Hensen′s node, zone of polarizing activity in the posterior limb bud, notochord, and floor plate of the neural tube. Downstream targets of Shh include the transcription factors Gli3, responsible for Greigs polycephalosyndactyly in humans, and Hoxd13, responsible for polysyndactyly.

Physical form

Solution in 0.01 M phosphte buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinyue Hu et al.
Gut microbes, 15(1), 2221093-2221093 (2023-06-07)
Epithelial ovarian cancer (EOC) is the most lethal gynecological cancer, which remains a threat to female health at all ages. Hypotheses for EOC development include the continuous presence of inflammation, in which microbiota and inflammatory cytokines participate in cancer-related signaling
Shih-Hung Yang et al.
Oncology, 85(2), 86-94 (2013-07-19)
We investigated the association of the hedgehog pathway with nuclear factor (NF)-κB and clinical outcomes in pancreatic cancer patients. We analyzed tissue samples for the expression of NF-κB (RelA/p65), sonic hedgehog (Shh) and glioma-associated oncogene homolog 1 (Gli1) by immunohistochemistry
Abu Shadat M Noman et al.
Therapeutic advances in medical oncology, 12, 1758835920911229-1758835920911229 (2020-03-25)
Sonic hedgehog (Shh) and Nrf2 play a critical role in chemotherapeutic resistance. These two genes have been found to be dysregulated in head and neck squamous cell carcinomas (HNSCC). The purpose of this study was to analyze the expression, function
Role of Sonic hedgehog signaling pathway in neuroblastoma development
Shahi MH et al
Biol. Med., 1, Rev2-Rev2 (2009)
Abu Shadat M Noman et al.
Therapeutic advances in medical oncology, 12, 1758835920911229-1758835920911229 (2020-03-25)
Sonic hedgehog (Shh) and Nrf2 play a critical role in chemotherapeutic resistance. These two genes have been found to be dysregulated in head and neck squamous cell carcinomas (HNSCC). The purpose of this study was to analyze the expression, function

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service