Skip to Content
Merck
All Photos(1)

Key Documents

S7199

Sigma-Aldrich

Monoclonal Anti-SERCA3 antibody produced in mouse

~1.0 mg/mL, clone PL/IM430, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ATP2A3, Anti-ATPase, Ca(2+)-Transporting, Ubiquitous, Anti-Sarcoplasmic reticulum Ca(2+)-ATPase 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PL/IM430, monoclonal

form

buffered aqueous solution

mol wt

antigen ~100 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 0.25-0.5 μg/mL using platelet total cell extract.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP2A3(489)

General description

Monoclonal Anti-SERCA3 (mouse IgG1 isotype) is derived from the hybridoma PL/IM430 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. The ATPase sarcoplasmatic/endoplasmatic reticulum Ca2+ transporting 3 (ATP2A3) gene codes for Ca2+-ATPase3 from the sarco/endoplasmic reticulum (SERCA3) and the coded protein is expressed in endothelial and epithelial tissues. The human SERCA3 gene gives rise to six 3¢-end splice variants encoding proteins varying in their C-terminal part and termed SERCA3a-f. SERCA3 gene is mapped to human chromosome 17p13.3.

Specificity

Monoclonal Anti-SERCA3 recognizes human SERCA3.

Application

Monoclonal Anti-SERCA3 antibody produced in mouse may be used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunocytochemistry
  • western blotting
  • immunoprecipitation
  • immunohistochemistry

Biochem/physiol Actions

Sarco/endoplasmic calcium ATPase pumps (SERCA) protein plays a pivotal role in intracellular calcium [Ca2+] homeostasis by lowering the cytoplasmic [Ca2+] concentration. Expression of sarco/endoplasmic reticulum (SERCA3) gene is reduced in gastric/colon carcinoma and leukemia cell types.The ATPase sarcoplasmatic/endoplasmatic reticulum Ca2+ transporting 3 (ATP2A3) gene may serve as an important target for salinomycin. Overexpression of this gene stimulates apoptosis. SERCA3 protein plays an important role in Ca2+ transport.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze at –20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aberrant SERCA3 expression during the colorectal adenoma-adenocarcinoma sequence
Gou WF, et al.
Oncology Reports, 31(1), 232-240 (2014)
Salinomycin triggers endoplasmic reticulum stress through ATP2A3 upregulation in PC-3 cells
Zhang Y, et al.
BMC Cancer, 19(1), 1-13 (2019)
Role of PKA in the process of neonatal cardiomyocyte hypertrophy induced by urotensin II
Xu J, et al.
International Journal of Molecular Medicine, 40(2), 499-504 (2017)
Drug Interactions With the Ca2+-ATPase From Sarco (Endo) Plasmic Reticulum (SERCA)
Tadini BF, et al.
Frontiers in molecular biosciences, 5(1), 36-36 (2018)
Jianrong Xu et al.
International journal of molecular medicine, 40(2), 499-504 (2017-06-29)
The model of urotensin II (UII)-induced cardiomyocyte hypertrophy has been widely used in studies on hypertrophy. However, the molecular mechanisms responsible for UII-induced cardiomyocyte hypertrophy have not yet been fully elucidated. It has been demonstrated that cardiomyocyte hypertrophy induced by UII

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service