S6821
Anti-SMC1L1 antibody produced in rabbit
IgG fraction of antiserum
Synonym(s):
Anti-SMC1, Anti-SMC1 structural maintenance of chromosomes 1-like 1, Anti-SMC1α
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
mol wt
antigen 160 kDa
species reactivity
human
technique(s)
immunoprecipitation (IP): 5-10 μL using 293-T cell lysates
western blot: 1:2,000-1:4,000 using extracts of HeLa nuclear cells
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... SMC1A(8243)
mouse ... Smc1a(24061)
rat ... Smc1a(63996)
General description
Specificity
Immunogen
Application
Biochem/physiol Actions
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service