Skip to Content
Merck
All Photos(1)

Key Documents

S6768

Sigma-Aldrich

N-Succinyl-Phe-Gly-Leu-β-naphthylamide

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$253.00

$253.00


Check Cart for Availability


Select a Size

Change View
50 MG
$253.00

About This Item

Empirical Formula (Hill Notation):
C31H36N4O6
CAS Number:
Molecular Weight:
560.64
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

$253.00


Check Cart for Availability

form

solid

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)CNC(=O)C(Cc1ccccc1)N2C(=O)CCC2=O)C(=O)Nc3ccc4ccccc4c3

InChI

1S/C31H34N4O5/c1-20(2)16-25(30(39)33-24-13-12-22-10-6-7-11-23(22)18-24)34-27(36)19-32-31(40)26(17-21-8-4-3-5-9-21)35-28(37)14-15-29(35)38/h3-13,18,20,25-26H,14-17,19H2,1-2H3,(H,32,40)(H,33,39)(H,34,36)

InChI key

SGVJLMFFXDTAFJ-UHFFFAOYSA-N

Substrates

Substrate for an LHRH-degrading neutral endopeptidase

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Horsthemke et al.
Biochemistry, 19(13), 2867-2873 (1980-06-24)
A neutral endopeptidase which degrades luteinizing hormone-releasing hormone (LH-RH, <GLu-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-GLy-NH2) has been purified 900-fold from extracts of bovine anterior pituitary. This Ca2+-independent enzyme of 83 000 molecular weight (as estimated by gel filtration) cleaves LH-RH (KM = 180 microM) at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service