Skip to Content
Merck
All Photos(1)

Documents

S6647

Sigma-Aldrich

[D-Pro4, D-Trp7,9, Nle11]-Substance P Fragment 4-11

≥95% (HPLC)

Synonym(s):

D-Pro-Gln-Gln-D-Trp-Phe-D-Trp-Leu-Nle-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H77N13O10
CAS Number:
Molecular Weight:
1116.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CCCCC(NC(=O)C(CC(C)C)NC(=O)C(Cc1c[nH]c2ccccc12)NC(=O)C(Cc3ccccc3)NC(=O)C(Cc4c[nH]c5ccccc45)NC(=O)C(CCC(N)=O)NC(=O)C(CCC(N)=O)NC(=O)C6CCCN6)C(N)=O

InChI

1S/C58H77N13O10/c1-4-5-18-41(51(61)74)65-55(78)45(27-33(2)3)68-57(80)48(30-36-32-64-40-20-12-10-17-38(36)40)71-56(79)46(28-34-14-7-6-8-15-34)69-58(81)47(29-35-31-63-39-19-11-9-16-37(35)39)70-54(77)44(23-25-50(60)73)67-53(76)43(22-24-49(59)72)66-52(75)42-21-13-26-62-42/h6-12,14-17,19-20,31-33,41-48,62-64H,4-5,13,18,21-30H2,1-3H3,(H2,59,72)(H2,60,73)(H2,61,74)(H,65,78)(H,66,75)(H,67,76)(H,68,80)(H,69,81)(H,70,77)(H,71,79)

InChI key

OFDJVNRFKDPBKR-UHFFFAOYSA-N

Gene Information

human ... TAC1(6863)

Biochem/physiol Actions

A potent substance P antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of substance P antagonists: [D-Pro4, D-Trp7,9,Nle11]-substance P(4-11).
G Köller et al.
Die Pharmazie, 39(1), 65-67 (1984-01-01)
G Bredy-Dobreva et al.
Acta physiologica et pharmacologica Bulgarica, 15(3), 10-15 (1989-01-01)
The effects of a new analogue of substance P (SP-A) and its interaction with substance P (SP) were studied in vitro isolated preparations from rabbit renal arteries and rat renal and pulmonary arteries. SP-A in did not affect the resting
Ruofan Wang et al.
Cell, 184(1), 106-119 (2020-12-18)
The Coronaviridae are a family of viruses that cause disease in humans ranging from mild respiratory infection to potentially lethal acute respiratory distress syndrome. Finding host factors common to multiple coronaviruses could facilitate the development of therapies to combat current

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service