Skip to Content
Merck
All Photos(2)

Key Documents

S5070

Sigma-Aldrich

Anti-Sodium Channel ASIC3 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-DRASIC

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
$398.00

$398.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
50 μL
$398.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$398.00


Estimated to ship on12 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200 using rat dorsal root ganglion lysate

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACCN3(9311)
mouse ... Accn3(171209)
rat ... Accn3(286920)

Immunogen

synthetic peptide corresponding to residues 2-16 of rat ASIC3. The sequence has 14/15 residues identical in mouse and 11/15 identical in human.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, 1% bovine serum albumin, and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service