Skip to Content
Merck
All Photos(1)

Key Documents

S4324

Millipore

Seppro® Stripping Buffer

Synonym(s):

glycine-based buffer

Sign Into View Organizational & Contract Pricing

Select a Size

200 ML
$179.00

$179.00


Available to ship on07 April 2025Details



Select a Size

Change View
200 ML
$179.00

About This Item

UNSPSC Code:
41106500
NACRES:
NA.32

$179.00


Available to ship on07 April 2025Details


storage temp.

2-8°C

General description

Seppro® stripping buffer is a glycine-based buffer (pH 2.25-2.45) recommended for use with Seppro® reagents and columns.

Application

Seppro® Stripping Buffer has been used:
  • for IgY14 and SuperMix depletion of human plasma[1]
  • immunoglobulin Y (IgY) immunodepletion of top abundant plasma proteins[2]
  • to strip the blots for western blotting[3]

Legal Information

Seppro is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Simon Sheng et al.
Methods in molecular biology (Clifton, N.J.), 728, 29-46 (2011-04-07)
Serum- and plasma-based biomarker discovery requires technologies with specific capabilities: sufficient proteome coverage and depth, technical reproducibly, and the scalability to enable analysis on a large number of samples at reasonable cost. We have shown that plasma samples processed using
Hasmik Keshishian et al.
Nature protocols, 12(8), 1683-1701 (2017-07-28)
Proteomic characterization of blood plasma is of central importance to clinical proteomics and particularly to biomarker discovery studies. The vast dynamic range and high complexity of the plasma proteome have, however, proven to be serious challenges and have often led
Anahid Ehteda et al.
BMC cancer, 13, 86-86 (2013-02-26)
Albendazole (ABZ) is a microtubule-targeting anthelmintic with a remarkable activity against a variety of human cancer cells. In this study, we examined if the antitumor activity of ABZ could be enhanced by its combination with other microtubule-binding agents. The interactions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service