Skip to Content
Merck
All Photos(1)

Key Documents

S3316

Sigma-Aldrich

SKF-89145 hydrobromide

>98% (HPLC), solid

Synonym(s):

4-(3,4-Dihydroxyphenyl)-6-methyl-4,5,6,7-tetrahydro­thieno[2,3-c]pyridine hydrobromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$229.00

$229.00


Check Cart for Availability
New, lower price on this item!


Select a Size

Change View
5 MG
$229.00

About This Item

Empirical Formula (Hill Notation):
C14H15NO2S · HBr
CAS Number:
Molecular Weight:
342.25
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77

$229.00


Check Cart for Availability
New, lower price on this item!

Quality Level

Assay

>98% (HPLC)

form

solid

color

off-white

solubility

DMSO: soluble 10 mg/mL
H2O: soluble 4 mg/mL

SMILES string

Br[H].CN1CC(c2ccc(O)c(O)c2)c3ccsc3C1

InChI

1S/C14H15NO2S.BrH/c1-15-7-11(10-4-5-18-14(10)8-15)9-2-3-12(16)13(17)6-9;/h2-6,11,16-17H,7-8H2,1H3;1H

InChI key

WUWJEHPRFDIUMV-UHFFFAOYSA-N

Application

SKF-89145 was used to treat Crassostrea virginica to study the role of D1 dopamine receptors in dopaminergic neurotoxicity of manganese in gills.[1]

Biochem/physiol Actions

D1 dopamine receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Anador et al.
In vivo, 32(3), 74-79 (2011-08-19)
The lateral epithelial cells of gill of Crassostrea virginica are innervated by dopamine and serotonin nerves that regulate the beating rate of their lateral cilia. Terminal release of dopamine slows down the beating rate of the cilia, while serotonin release

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service