Skip to Content
Merck
All Photos(1)

Documents

S3071

Sigma-Aldrich

γ-Synuclein human

recombinant, expressed in E. coli, N-terminal histidine tagged, ≥90% (SDS-PAGE), lyophilized powder

Synonym(s):

BCSG1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

Assay

≥90% (SDS-PAGE)

form

lyophilized powder

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SNCG(6623)

Biochem/physiol Actions

13.3 kDa (127 amino acids) protein, which plays a role in neurofilament network integrity. Member of a family of small, highly conserved synuclein proteins expressed primarily in neurons and in certain tumors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

γ-Synuclein: subcellular localization in neuronal and non-neuronal cells and effect on signal transduction.
Surguchov, A, et al.
Cell Motility, 49, 218-222 (2001)
Zhong-Zong Pan et al.
The Journal of biological chemistry, 277(38), 35050-35060 (2002-07-18)
Synucleins are a family of highly conserved small proteins predominantly expressed in neurons. Recently we and others have found that gamma-synuclein is dramatically up-regulated in the vast majority of late-stage breast and ovarian cancers and that gamma-synuclein over-expression can enhance
Wei Wang et al.
BMC biochemistry, 12, 63-63 (2011-12-14)
D-serine is an important coagonist at the NR1 subunit of the NMDA receptor class of glutamate receptors. It is chiefly synthesized in the CNS by serine racemase (SR). Regulation of SR activity is still poorly understood. As step toward developing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service