Skip to Content
Merck
All Photos(1)

Key Documents

S3000

Sigma-Aldrich

Dimethyl sebacate

≥95%

Synonym(s):

Decanedioic acid 1,10-dimethyl ester, Dimethyl decanedioate, NSC 9415, Sebacic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCO(CH2)8COOCH3
CAS Number:
Molecular Weight:
230.30
Beilstein:
1785523
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥95%

bp

158 °C/10 mmHg (lit.)

mp

29-31 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

functional group

ester

shipped in

ambient

storage temp.

room temp

SMILES string

COC(=O)CCCCCCCCC(=O)OC

InChI

1S/C12H22O4/c1-15-11(13)9-7-5-3-4-6-8-10-12(14)16-2/h3-10H2,1-2H3

InChI key

ALOUNLDAKADEEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Determination of dimethylacrylic acid and its ethyl ester in the air of a work area].
N Ia Smoliar
Gigiena i sanitariia, (1)(1), 51-52 (1984-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service