S1818
SNX-482 synthetic
>98% (HPLC), lyophilized powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
Recommended Products
Assay
>98% (HPLC)
form
lyophilized powder
mol wt
apparent mol wt ~4.5 kDa by SDS-PAGE
solubility
aqueous buffer: soluble
storage temp.
−20°C
Amino Acid Sequence
Gly-Va-lAsp-Lys-Ala-Gly-Cys-Arg-Tyr-Met-Phe-Gly-Gly-Cys-Ser-Val-Asn-Asp-Asp-Cys-Cys-Pro-Arg-Leu-Gly-Cys-His-Ser-Leu-Phe-Ser-Tyr-Cys-Ala-Trp-Asp-Leu-Thr-Phe-Ser-Asp
Biochem/physiol Actions
Specifically blocks Cav2.3 (α1E, R-type) calcium channels in a voltage dependent manner.
Specifically blocks Cav2.3 (α1E, R-type) calcium channels.
Recommended products
Recombinant SNX-482 is also available from Sigma. To learn more about and purchase recombinant SNX-482, click here.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service