S1772
Sodium chloride solution 15%
150 g/L sodium chloride in H2O, tissue culture grade, suitable for insect cell culture
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161700
Recommended Products
sterility
sterile-filtered
concentration
150 g/L sodium chloride in H2O, tissue culture grade
technique(s)
cell culture | insect: suitable
impurities
endotoxin, tested
storage temp.
−20°C
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service