Skip to Content
Merck
All Photos(1)

Key Documents

S1688

Sigma-Aldrich

SR 57227A

solid

Synonym(s):

4-amino-1-(6-chloro-2-pyridyl)-piperidine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$170.00

$170.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$170.00

About This Item

Empirical Formula (Hill Notation):
C10H14ClN3 · HCl
CAS Number:
Molecular Weight:
248.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$170.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

form

solid

color

white

solubility

H2O: soluble 9 mg/mL

SMILES string

Cl.NC1CCN(CC1)c2cccc(Cl)n2

InChI

1S/C10H14ClN3.ClH/c11-9-2-1-3-10(13-9)14-6-4-8(12)5-7-14;/h1-3,8H,4-7,12H2;1H

InChI key

FUMINTAAUJUVMP-UHFFFAOYSA-N

Application

SR 57227A may be used in 5-HT3-mediated cell signaling studies.

Biochem/physiol Actions

Potent and selective 5-HT3 serotonin receptor agonist.
SR 57227A is a highly selective of central and peripheral 5-HT3 receptors[1] and an inhibitor of NMDA receptor-mediated responses in rat cortical pyramidal cells.[2] It exhibits anti-depressant like effects in rats and decreased isolation-induced aggressive behavior.[3]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiao Jun Zhang et al.
Brain research, 1384, 69-79 (2011-02-05)
In the present study, effect of SR 57227A, a selective 5-hydroxytryptamine-3 (5-HT(3)) receptor agonist, on the firing activity of pyramidal neurons in the medial prefrontal cortex (mPFC) was studied in normal rats and rats with 6-hydroxydopamine lesions of the substantia
Taha Gholipour et al.
Seizure, 19(1), 17-22 (2009-11-28)
There is some evidence that epileptic seizures could be induced or increased by 5-hydroxytryptamine (5-HT) attenuation, while augmentation of serotonin functions within the brain (e.g. by SSRIs) has been reported to be anticonvulsant. This study was performed to determine the
X Liang et al.
Synapse (New York, N.Y.), 29(3), 257-268 (1998-06-23)
The techniques of intracellular recording and single-electrode voltage-clamp were used to study the effect of serotonin (5-HT) and the selective 5-HT3 receptor agonist SR 57227A on N-methyl-D-aspartic acid (NMDA)-evoked responses in pyramidal cells of the rat medial prefrontal cortex (mPFC)
Pierre A Guertin et al.
Journal of neurophysiology, 94(5), 3397-3405 (2005-07-29)
Centrally expressed 5-HT3 receptors (5-HTR3) are well known for their role in wakefulness, cognition, and nociception. However, clear evidence of their participation in motor control is still lacking despite specific 5-HTR3 expression in hindlimb motor areas of the spinal cord
Marlies Oostland et al.
The Journal of physiology, 589(Pt 20), 4837-4846 (2011-09-01)
The serotonin 5-HT(3) receptor is the only ligand-gated ion channel activated by serotonin and is expressed by GABAergic interneurons in many brain regions, including the cortex, amygdala and hippocampus. Furthermore, 5-HT(3) receptors are expressed by glutamatergic Cajal-Retzius cells in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service