Skip to Content
Merck
All Photos(1)

Key Documents

S1300

Sigma-Aldrich

Monoclonal Anti-L-Selectin (CD62L) antibody produced in mouse

2 mg/mL, clone FMC46, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-CD62L, Anti-L-Selectin

Sign Into View Organizational & Contract Pricing

Select a Size

0.2 ML
$572.00

$572.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.2 ML
$572.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$572.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

FMC46, monoclonal

form

buffered aqueous solution

mol wt

antigen 74-95 kDa

species reactivity

human

concentration

2 mg/mL

technique(s)

flow cytometry: suitable
immunohistochemistry (frozen sections): suitable

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SELL(6402)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Specificity

Recognizes the CD62L (L-Selectin, LAM-1, Leu8, TQ1, LECAM-1, LECCAM-1, DREG) surface antigen. The antibody recognizes an epitope localized to the lectin domain. It inhibits "rolling" interactions and significantly reduces the binding of FITC-yeast phosphomannan monoester core polysaccharide to lymphocytes, monocytes and neutrophils.
5th Workshop: Code No. S061

Immunogen

human PHA-activated peripheral blood mononuclear cells.

Target description

CD62L, a glycoprotein and member of the selectin family of adhesion receptors, is constitutively expressed on all classes of leukocytes including lymphocytes, monocytes, and polymorphonuclear cells. It is expressed on myeloid progenitor cells, erythroid precursor cells, and some thymocytes.

Physical form

Solution, 0.2 μm filtered, in 0.01 M phosphate buffered saline, pH 7.4.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service