Skip to Content
Merck
All Photos(1)

Documents

S104

Supelco

(−)-Scopolamine hydrobromide trihydrate

Synonym(s):

Hyoscine hydrobromide, Scopine tropate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO4 · HBr · 3H2O
CAS Number:
Molecular Weight:
438.31
Beilstein:
6100669
MDL number:
UNSPSC Code:
41116107

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

195-199 °C (dry matter) (lit.)

format

neat

SMILES string

O.O.O.Br.CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4

InChI

1S/C17H21NO4.BrH.3H2O/c1-18-13-7-11(8-14(18)16-15(13)22-16)21-17(20)12(9-19)10-5-3-2-4-6-10;;;;/h2-6,11-16,19H,7-9H2,1H3;1H;3*1H2/t11-,12-,13-,14+,15-,16+;;;;/m1..../s1

InChI key

LACQPOBCQQPVIT-SEYKEWMNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Competitive nonselective muscarinic acetylcholine antagonist. Scopolamine-induced amnesia in laboratory animals is a commonly-used model of memory deficit.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Reiss et al.
Psychopharmacology, 231(22), 4337-4347 (2014-04-29)
Recognition memory is an important aspect of human declarative memory and is one of the routine memory abilities altered in patients with amnestic syndrome and Alzheimer's disease. In rodents, recognition memory has been most widely assessed using the novel object
Han Jin Oh et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 30(7), 533-542 (2014-04-29)
To investigate the efficacy of hypotonic 0.18% sodium hyaluronate (SH) eyedrops in a mouse model of experimental dry eye (EDE). EDE was induced in C57BL/6 mice by a subcutaneous scopolamine injection and an air draft. The mice were divided into
Natalija Popović et al.
PloS one, 9(6), e100348-e100348 (2014-06-18)
The effects of scopolamine on memory consolidation are controversial and depend on several factors (i.e. site of administration, time of administration and testing, dose, cognitive task, experimental protocol, specie, strain, etc.). Generally, the range dose of systemic administered scopolamine, used
Zhengri Li et al.
Current eye research, 39(9), 871-878 (2014-02-25)
To investigate the efficacy of the topical application of omega-3 essential fatty acids (EFAs) and hyaluronic acid (HA) mixtures in a mouse model of experimental dry eye (EDE). Eye drops consisting of 0.1% HA, 0.02%, or 0.2% omega-3 EFAs alone
Sasha Devore et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(34), 11244-11260 (2014-08-22)
The olfactory bulb (OB) and piriform cortex receive dense cholinergic projections from the basal forebrain. Cholinergic modulation within the piriform cortex has long been proposed to serve important functions in olfactory learning and memory. We here investigate how olfactory discrimination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service