Skip to Content
Merck
All Photos(2)

Key Documents

S0191

Sigma-Aldrich

Sonic Hedgehog Peptide from mouse

≥97% (SDS-PAGE), recombinant, expressed in E. coli, Fused to a histidine tag at the C-terminus, lyophilized powder

Synonym(s):

Shh

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$591.00

$591.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$591.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

$591.00


Estimated to ship on26 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

0.4-3.0 μg/mL ED50

mol wt

predicted mol wt 20 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Shh(20423)

General description

Sonic hedgehog (Shh) is a morphogenic glycoprotein. The gene encoding it is localized on mouse chromosome 5.[1][2]

Application

Sonic hedgehog peptide from mouse has been used for neural differentiation.[3]

Biochem/physiol Actions

Cell signaling peptide involved in embryonic morphogenic development of several tissues including Hensen′s node, the notochord, and the neural tube.
Involved in embryonic morphogenic development of several tissues including Hensen′s node, the notochord, and the neural tube.
Sonic hedgehog (Shh) has a role in fracture healing and regeneration.[1] The protein associates with its receptor Patched (Ptc-1), de-represses Smoothened (Smo) and activates the Gli family of transcription factors.[4] Mutations in the Shh pathway have been linked to developmental diseases and cancer.[5]

Other Notes

N-terminal peptide encoding amino acid residues 25-198.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline, pH 7.4, containing 5% trehalose and 50 μg of bovine serum albumin per 1 μg of cytokine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na Song et al.
Experimental and therapeutic medicine, 15(6), 5084-5090 (2018-05-29)
Diabetes mellitus (DM) complications affect patients and cause varying damage. Skin ulcers exhibit difficulties in wound healing, and the regulatory basis for this remains unclear. High glucose concentration (HG) was utilized to mimic DM in cultured cells. Reverse transcription-quantitative polymerase
Abolghasem Esmaeili et al.
Avicenna journal of medical biotechnology, 6(1), 21-26 (2014-02-20)
Stem cells from Human Exfoliated Deciduous teeth (SHED) have the capability to differentiate into neural cells. Neurotrophins including Nerve Growth Factor (NGF), Brain-Derived Neurotrophic Factor (BDNF), neurotrophin-3 (NT-3), and neurotrophin-4 (NT-4) have neurogenesis, neurotrophic, or neuroprotective effects and are expressed
Sanming Li et al.
Journal of cellular and molecular medicine, 22(1), 230-240 (2017-08-08)
Ectodysplasin A (Eda), a member of the tumour necrosis factor superfamily, plays an important role in ectodermal organ development. An EDA mutation underlies the most common of ectodermal dysplasias, that is X-linked hypohidrotic ectodermal dysplasia (XLHED) in humans. Even though
Cloning, expression, and chromosomal location of SHH and IHH: two human homologues of the Drosophila segment polarity gene hedgehog.
Marigo V
Genomics, 28(1), 44-51 (1995)
Sonic hedgehog stimulates neurite outgrowth in a mechanical stretch model of reactive-astrogliosis.
Berretta A
Scientific Reports, 6, 21896-21896 (2016)

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service