Skip to Content
Merck
All Photos(1)

Key Documents

RES7102I

SAFC

Imidazole hydrochloride

Synonym(s):

Imidazole hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2 · HCl
CAS Number:
Molecular Weight:
104.54
Beilstein:
3678283
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21
Pricing and availability is not currently available.

mp

158-161 °C (lit.)

SMILES string

Cl.c1c[nH]cn1

InChI

1S/C3H4N2.ClH/c1-2-5-3-4-1;/h1-3H,(H,4,5);1H

InChI key

JDIIGWSSTNUWGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service