Skip to Content
Merck
All Photos(1)

Key Documents

RES2029D-A7

SAFC

Dextran sulfate sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$508.00
100 G
$2,479.00
1 KG
$9,301.00

$508.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$508.00
100 G
$2,479.00
1 KG
$9,301.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.21

$508.00


Check Cart for Availability

Request a Bulk Order

biological source

synthetic

Quality Level

form

powder

optical activity

[α]20/D 75 to 105 °

mol wt

Mw 8000 g/mol

technique(s)

cell culture | mammalian: suitable

impurities

Free Inorganic Sulfate; tested, Chloride; tested

suitability

suitable for manufacturing use

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to documentation for this product requires a confidentiality disclosure agreement.

Packaging

Product is available in the following package sizes:
RES2029D-A701X: 100 g container
RES2029D-A702X: 1 kg container
RES2029D-A707X: 10 g container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service