Skip to Content
Merck
All Photos(4)

Key Documents

RDD017

Sigma-Aldrich

Ethylenediaminetetraacetic acid

anhydrous, free-flowing, powder, Redi-Dri, ACS reagent, 99.4-100.6%

Synonym(s):

(Ethylenedinitrilo)tetraacetic acid, EDTA, Edathamil, Ethylenedinitrilotetraacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$199.00
1 KG
$326.00

$199.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$199.00
1 KG
$326.00

About This Item

Linear Formula:
(HO2CCH2)2NCH2CH2N(CH2CO2H)2
CAS Number:
Molecular Weight:
292.24
Beilstein:
1716295
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

$199.00


Check Cart for Availability

Request a Bulk Order

grade

ACS reagent

Quality Level

product line

Redi-Dri

Assay

99.4-100.6%

form

powder

quality

anhydrous
free-flowing

reaction suitability

reagent type: chelator

technique(s)

cell culture | mammalian: suitable

impurities

≤0.005% Insol. Dil. NH4OH
≤0.1% Nitrilotriacetic acid [[HOCOCH23N]

ign. residue

≤0.2%

pH

2.5 (23 °C, 10 g/L)

mp

250 °C (dec.) (lit.)

solubility

H2O: soluble 0.4 g/L at 20 °C

density

1.46 g/cm3 at 20 °C

cation traces

Ca: ≤0.001%
Fe: ≤0.005%
Mg: ≤5 ppm
heavy metals (as Pb): ≤0.001%

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)

InChI key

KCXVZYZYPLLWCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service