Skip to Content
Merck
All Photos(4)

Key Documents

RDD015

Sigma-Aldrich

CAPS

anhydrous, free-flowing, Redi-Dri, ≥99%

Synonym(s):

3-(Cyclohexylamino)-1-propanesulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$550.00
1 KG
$1,010.00

$550.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$550.00
1 KG
$1,010.00

About This Item

Linear Formula:
C6H11NH(CH2)3SO3H
CAS Number:
Molecular Weight:
221.32
Beilstein:
2835588
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

$550.00


Check Cart for Availability

Request a Bulk Order

grade

anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

impurities

≤1% water (Karl Fischer)

pH

3.0-7.0

pKa 

10.4

mp

>300 °C (lit.)

solubility

water: 15 % (w/w), clear, colorless

absorption

≤0.05 at 290 nm at 15 % (w/w)

SMILES string

OS(=O)(=O)CCCNC1CCCCC1

InChI

1S/C9H19NO3S/c11-14(12,13)8-4-7-10-9-5-2-1-3-6-9/h9-10H,1-8H2,(H,11,12,13)

InChI key

PJWWRFATQTVXHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service