Skip to Content
Merck

Skip To

RAB1663

Human Renin ELISA

for serum, plasma and cell culture supernatants

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Check Cart for Availability
$1,060.00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... REN(5972)
Input:
sample type plasma
sample type cell culture supernatant(s)
sample type serum
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

$1,060.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.41 ng/mL
standard curve range: 0.410-100 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... REN(5972)

General description

This ELISA antibody pair detects Human Renin (REN)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0565RAB1438RAB1581
species reactivity

human

species reactivity

mouse

species reactivity

human

species reactivity

human

Gene Information

human ... REN(5972)

Gene Information

mouse ... Ren1(19701)

Gene Information

human ... EPHA2(1969)

Gene Information

human ... MEPE(56955)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.41 ng/mL
standard curve range: 0.410-100 ng/mL

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 6 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.08 ng/mL
standard curve range: 0.082-20 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8.4 ng/mL
standard curve range: 8.192-2000 ng/mL

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters