Skip to Content
Merck
All Photos(1)

Key Documents

RAB1351

Sigma-Aldrich

Human CD74 ELISA

for serum, plasma and cell culture supernatants

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$1,150.00

$1,150.00


Estimated to ship on04 May 2025



Select a Size

Change View
96 WELLS
$1,150.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$1,150.00


Estimated to ship on04 May 2025


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.1 ng/mL
standard curve range: 0.102-25 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CD74(972)

General description

The CD74 (HLA-DR (human leukocyte antigen – antigen D related) antigens-associated invariant chain) gene encodes for a type II transmembrane protein and is expressed by antigen presenting cells such as monocytes, B cells, macrophages and dendritic cells.[1][2] The CD74 gene is mapped to human chromosome 5q33.1.[3]

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

CD74 (HLA-DR (human leukocyte antigen – antigen D related) antigens-associated invariant chain) is part of major histocompatibility complex (MHC) class II and its function is associated with antigen presentation. Upregulation of the CD74 gene is observed cancer including breast cancer and it is known to promote to metastasis.[4] CD74 also serves as a cytokine receptor in immune and epithelial cells. It induces cellular processes such as cell proliferation, migration and survival pathways.[5]

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Van?den Berghe?s 5q?syndrome in 2008.
Mohamedali A and Mufti G J
British Journal of Haematology, 144(2), 157-168 (2009)
CD74 and intratumoral immune response in breast cancer.
Wang Z Q, et al.
Oncotarget, 8(8), 12664-12664 (2017)
GSK-3?-mediated fatty acid synthesis enhances epithelial to mesenchymal transition of TLR4-activated colorectal cancer cells through regulation of TAp63.
Park G B, et al.
International Journal of Oncology, 49(5), 2163-2172 (2016)
Gabriela Athziri Sánchez-Zuno et al.
Journal of clinical medicine, 11(1) (2022-01-12)
Macrophage migration inhibitory factor (MIF) significantly contributes to rheumatoid arthritis (RA) pathogenesis. We aimed to evaluate the canonical (CD74/CD44) and non-canonical MIF receptors (CXCR2,4 and 7) expression and sCD74 to establish their association with RA clinical activity according to DAS28-ESR.
Modeling of both shared and distinct interactions between MIF and its homologue D-DT with their common receptor CD74.
Meza-Romero R, et al.
Cytokine, 88, 62-70 (2016)

Questions

  1. Hi, can you tell me what are the volume of all reagents at Human CD74 ELISA (RAB1351)? Thanks a lot!

    1 answer
    1. Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service