Skip to Content
Merck
All Photos(1)

Key Documents

RAB1178

Sigma-Aldrich

Human MMR ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$1,120.00

$1,120.00


Check Cart for Availability


Select a Size

Change View
96 WELLS
$1,120.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$1,120.00


Check Cart for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum sample(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.41 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MRC1(4360)

General description

This ELISA antibody pair detects Human Macrophage Mannose Receptor (MRC1 / CD206)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya-Wen Shen et al.
Mediators of inflammation, 2020, 7948095-7948095 (2020-11-17)
Soluble CD206 (sCD206) is considered a macrophage activation marker, and a previous study proved it as a potential biomarker to predict the severity of anti-melanoma differentiation-associated gene 5- (anti-MDA-5-) positive dermatomyositis- (DM-) associated interstitial lung disease (ILD). To investigate the
Dania AlQasrawi et al.
Microorganisms, 8(5) (2020-05-07)
Cigarette smoke (CS) exacerbates symptoms in Crohn's disease (CD) patients while protecting those with ulcerative colitis (UC). CD has been associated with immuno-dysregulation, mucosal dysfunction, and infection. Among the CD-debated pathogens are Mycobacterium avium&nbsp;subsp.&nbsp;paratuberculosis (MAP), adherent invasive Escherichia coli (AIEC)
Sudhir K Yadav et al.
Journal of clinical medicine, 10(4) (2021-03-07)
Dimethyl fumarate (DMF) is an oral agent for relapsing-remitting multiple sclerosis (RRMS). In this study, we investigated the therapeutic mechanism of DMF using experimental autoimmune encephalomyelitis (EAE). DMF treatment decreased the proliferation of T cells and the production of IL-17A
Banishree Saha et al.
Hepatology (Baltimore, Md.), 70(4), 1134-1149 (2019-03-21)
Although mortality due to acute alcoholic hepatitis (AH) correlates with Model for End-Stage Liver Disease (MELD) scores, biomarkers are critically needed to manage this disease. Increases in inflammatory markers and macrophage activation are associated with acute AH and could be

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service