Skip to Content
Merck
All Photos(1)

Key Documents

RAB1097

Sigma-Aldrich

Human Vitronectin ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$970.00

$970.00


Estimated to ship on26 May 2025



Select a Size

Change View
96 WELLS
$970.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$970.00


Estimated to ship on26 May 2025


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum sample(s)
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.8 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VTN(7448)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnus Paulsson et al.
Frontiers in microbiology, 9, 1559-1559 (2018-08-01)
Pathogens causing pneumonia utilize the complement regulator vitronectin to evade complement-mediated killing. Although vitronectin is associated with several chronic lung diseases, the role of bronchoalveolar vitronectin in pneumonia has not been studied. This study sought to reveal the involvement of
M Luo et al.
Journal of thrombosis and haemostasis : JTH, 15(12), 2451-2460 (2017-10-14)
Essentials Vitronectin (VN) is produced by smooth muscle cells (SMCs) and promotes neointima formation. We studied the regulation of vascular VN expression by plasminogen activator inhibitor-1 (PAI-1). PAI-1 stimulates VN gene expression in SMCs by binding LDL receptor-related protein 1.
Petra M Schulz et al.
Biologicals : journal of the International Association of Biological Standardization, 52, 72-77 (2018-01-18)
Fibryga® is a new lyophilized fibrinogen concentrate for intravenous use for the treatment of congenital fibrinogen deficiency. fibryga® is produced from pooled human plasma and the final product is characterized by high purity, integrity, and pathogen safety. Functional activity of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service