Skip to Content
Merck
All Photos(1)

Key Documents

RAB0960

Sigma-Aldrich

Phosphotyrosine FER / Tyrosine-protein Kinase Fer ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$1,500.00

$1,500.00


Estimated to ship on30 April 2025



Select a Size

Change View
96 WELLS
$1,500.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$1,500.00


Estimated to ship on30 April 2025


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type: human cell lysate

assay range

inter-assay cv: <10%
intra-assay cv: <12%

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FER(2241)

General description

FER (fps/fes related) tyrosine kinase is encoded by the gene mapped to human chromosome 5q21.3.[1] Tyrosine kinase is a cytoplasmic protein characterized with a single SH2 domain and an extensive amino terminus. The encoded protein is ubiquitously expressed.[2]
The antibody pair provided in this kit recognizes Human Tyrosine-Phosphorylated-FER.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

FER tyrosine kinase induces signal transduction at the plasma membrane.[3] It is implicated in the regulation of cell-cell adhesion.[4] FER tyrosine kinase interacts with phosphatidic acid (PA) to enhance phosphorylation of cortactin, a protein that promotes actin.[5]

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABHRP6Phosphotyrosine ELISA HRP-StreptavidinSDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of a de novo balanced translocation in a patient with moderate mental retardation and dysmorphic features
Haddad M R, et al.
European Journal of Medical Genetics, 52(4), 211-217 (2009)
The Tyrosine Kinase Fer Is a Downstream Target of the PLD-PA Pathway that Regulates Cell Migration
Itoh T, et al.
Science Signaling, 2(87), ra52-ra52 (2009)
The cytoplasmic tyrosine kinase FER is associated with the catenin-like substrate pp120 and is activated by growth factors.
Kim L and Wong T W
Molecular Cell Biology, 15(8), 4553-4561 (1995)
A curvature-dependent membrane binding by tyrosine kinase Fer involves an intrinsically disordered region.
Yamamoto H, et al.
Biochemical and Biophysical Research Communications, 495(1), 1522-1527 (2018)
Naoko Kogata et al.
Molecular biology of the cell, 14(9), 3553-3564 (2003-09-16)
Platelet endothelial adhesion molecule-1 (PECAM-1) is a part of intercellular junctions and triggers intracellular signaling cascades upon homophilic binding. The intracellular domain of PECAM-1 is tyrosine phosphorylated upon homophilic engagement. However, it remains unclear which tyrosine kinase phosphorylates PECAM-1. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service