Skip to Content
Merck
All Photos(1)

Documents

RAB0908

Sigma-Aldrich

Rat Mag / Myelin-associated Glycoprotein ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 10.24-2500 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Mag(29409)

General description

The antibody pair provided in this kit recognizes rat MAG / Myelin-associated Glycoprotein / Siglec-4a.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Myelin-associated glycoprotein (MAG) is expressed in Schwann cells and oligodendrocytes. It inhibits the growth of axons. MAG also has a role in signal transduction pathways and cell adhesion. In complex with the neurite outgrowth inhibitor (Nogo) receptor, MAG acts on neurons via the p75 receptor. It activates and modulates the function of Ras homolog gene family, member A (RhoA).

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Myelin-associated glycoprotein inhibits the neuronal differentiation of neural progenitors.
Li X, et al.
Neuroreport, 20(7), 708-712 (2009)
Myelin-associated glycoprotein inhibits microtubule assembly by a Rho-kinase-dependent mechanism.
Mimura F, et al.
The Journal of Biological Chemistry, 281(23), 15970-15979 (2006)
Tissue sparing and functional recovery following experimental traumatic brain injury is provided by treatment with an anti-myelin-associated glycoprotein antibody.
Thompson HJ, et al.
The European Journal of Neuroscience, 24(11), 3063-3072 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service