Skip to Content
Merck
All Photos(1)

Key Documents

RAB0785

Sigma-Aldrich

Mouse Fas L ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$970.00

$970.00


Estimated to ship on02 June 2025



Select a Size

Change View
96 WELLS
$970.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$970.00


Estimated to ship on02 June 2025


species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8 pg/mL
standard curve range: 8.19-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... FASLG(14103)

General description

The antibody pair provided in this kit recognizes mouse Fas Ligand.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ignatios Ikonomidis et al.
Circulation. Cardiovascular imaging, 7(4), 619-628 (2014-05-02)
We investigated the effects of anakinra, an interleukin-1 receptor antagonist, on coronary and left ventricular function in coronary artery disease (CAD) patients with rheumatoid arthritis. In a double-blind crossover trial, 80 patients with rheumatoid arthritis (60 with CAD and 20
Mark T Kearns et al.
American journal of physiology. Lung cellular and molecular physiology, 307(1), L62-L70 (2014-05-20)
Apoptosis of alveolar macrophages and their subsequent clearance by neighboring phagocytes are necessary steps in the resolution of acute pulmonary inflammation. We have recently identified that activation of the Fas death receptor on the cell surface of macrophages drives macrophage
Anil K Taneja et al.
International journal of cardiology, 175(2), 253-260 (2014-06-01)
Heart failure activates neurohormones, and elevated levels of brain natriuretic peptide (BNP) are associated with adverse outcomes. The SENIORS trial showed that nebivolol, a highly selective beta-1 antagonist with vasodilating properties, reduced the composite outcome of all cause mortality or
Soon Woo Nam et al.
Clinical science (London, England : 1979), 127(8), 519-526 (2014-04-10)
Cirrhotic cardiomyopathy is defined as systolic and diastolic dysfunctions, electrophysiological changes and macroscopic structural changes. However, the underlying mechanisms of this syndrome remain unclear. A possible role of myocardial apoptosis in the pathogenesis has not been previously examined. We hypothesized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service