Skip to Content
Merck
All Photos(1)

Key Documents

RAB0741

Sigma-Aldrich

Human THBS2 / Thrombospondin-2 ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$1,500.00

$1,500.00


Check Cart for Availability


Select a Size

Change View
96 WELLS
$1,500.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

$1,500.00


Check Cart for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.68 ng/mL
standard curve range: 0.655-160 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... THBS2(7058)

General description

The antibody pair provided in this kit recognizes Human Thrombospondin 2.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas W Berger et al.
Theranostics, 9(5), 1280-1287 (2019-03-15)
The most frequent malignancy of the pancreas is the pancreatic ductal adenocarcinoma (PDAC). Despite many efforts PDAC has still a dismal prognosis. Biomarkers for early disease stage diagnosis as a prerequisite for a potentially curative treatment are still missing. Novel
Shirsa Udgata et al.
Cancer prevention research (Philadelphia, Pa.), 14(2), 223-232 (2020-10-18)
Pancreatic ductal adenocarcinoma (PDAC) is often diagnosed too late for effective therapy. The classic strategy for early detection biomarker advancement consists of initial retrospective phases of discovery and validation with tissue samples taken from individuals diagnosed with disease, compared with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service