Skip to Content
Merck
All Photos(1)

Key Documents

RAB0709

Sigma-Aldrich

Human LHB / Lutropin Subunit Beta ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$970.00

$970.00


Estimated to ship on18 May 2025



Select a Size

Change View
96 WELLS
$970.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$970.00


Estimated to ship on18 May 2025


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 140 pg/mL
standard curve range: 156.3-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LHB(3972)

General description

The antibody pair provided in this kit recognizes human Lutropin Subunit Beta.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gervette M Penny et al.
Reproduction (Cambridge, England), 154(4), 455-467 (2017-07-16)
Testicular Leydig cells produce androgens essential for proper male reproductive development and fertility. Here, we describe a new Leydig cell ablation model based on Cre/Lox recombination of mouse
Weidong Zhang et al.
Chemosphere, 194, 147-157 (2017-12-05)
Numerous epidemiological studies suggest that air pollutants cause a decline in the quality of human spermatozoa and thus a reduction in fertility. However, the exact cause of infertility remains unknown. Air pollution gases, such as NH
Tiantian Song et al.
Frontiers in physiology, 8, 911-911 (2017-11-29)
The regulatory factors for stem Leydig cell development are largely unknown. Herein, we reported that parathyroid hormone-related protein (PTHrP) may be a factor to regulate this process. The effects of PTHrP on rat stem Leydig cell proliferation and differentiation were
Fereshteh Dardmeh et al.
PloS one, 12(10), e0185964-e0185964 (2017-10-11)
Probiotics have been proposed as alternatives to pharmacological products in several medical conditions including the modulation of obesity, which is frequently associated with poor semen quality. However, effects of probiotics on male fertility have been less investigated. This study assessed
Marie Alfrede Mvondo et al.
BMC complementary medicine and therapies, 20(1), 275-275 (2020-09-13)
Myrianthus arboreus P. Beauv. (Cecropiaceae) is a medicinal plant used to treat female infertility. The aqueous extract of M. arboreus leaves was found to improve the fertility of healthy female Wistar rats. In the present study, we proposed evaluating the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service