Skip to Content
Merck
All Photos(1)

Documents

RAB0689

Sigma-Aldrich

Human IL36G / Interleukin-36 Gamma ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 13 pg/mL
standard curve range: 12.29-3000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL36G(56300)

General description

The IL36G ELISA kit provides for the Quantitative measurement of IL-36 gamma in Cell Culture Supernatants, Plasma and Serum.
Interleukin-36 gamma (IL36G), also known as IL-1F9, is a member of interleukin (IL)-1-like proinflammatory cytokine family. It is encoded by the gene mapped to human chromosome 2q12-q21. IL36G is abundantly expressed in epithelial tissues and numerous myeloid-derived cell types.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Members of interleukin (IL)-1 family of cytokines play a vital role in regulating immune responses to infectious challenges and sterile insults. IL-36 is implicated in mucosal immunity, such as lung anti-bacterial responses. Interleukin-36 γ (IL36G) participates in inflammation response and differentiation. Overexpression of IL36G has been observed in psoriasis patients. Thus, down regulation IL36G can be considered as an effective method to treat psoriasis.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Participation of interleukin family 1 in the development of inflammatory reaction in the infectious process. 1. General characteristics of the interleukin family 1.
Abaturov AE, et al.
CHILDS HEALTH (2014)
IL-36γ inhibits differentiation and induces inflammation of keratinocyte via Wnt signaling pathway in psoriasis.
Wang W, et al.
International Journal of Medical Sciences, 14(10), 1002-1002 (2017)
IL-1 family cytokines in cardiovascular disease.
Pfeiler S, et al.
Cytokine (2017)
IL-36γ is a crucial proximal component of protective type-1-mediated lung mucosal immunity in Gram-positive and-negative bacterial pneumonia.
Kovach MA, et al.
Mucosal Immunology, 10(5), 1320-1320 (2017)
Jennifer Mildenberger et al.
Autophagy, 13(10), 1664-1678 (2017-08-19)
Inflammation is crucial in the defense against infections but must be tightly controlled to limit detrimental hyperactivation. Our diet influences inflammatory processes and omega-3 polyunsaturated fatty acids (n-3 PUFAs) have known anti-inflammatory effects. The balance of pro- and anti-inflammatory processes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service