species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
assay range
inter-assay cv: <10%
intra-assay cv: <12%
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CALB1(793)
General description
The CALB1 ELISA kit provides for the Quantitative measurement of Calbindin in Cell Culture Supernatants, Plasma and Serum.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of cellular biochemistry, 119(10), 8035-8047 (2018-01-30)
Calcium binding protein calbindin-D28K (CaBP28K) mediates the relationship between vitamin D and calcium, but its mechanism remains unclear during bone formation. The present study reports that maternal CaBP28K levels were positively correlated with paired umbilical cord CaBP28K levels. In addition
Journal of thrombosis and haemostasis : JTH, 12(7), 1066-1075 (2014-04-23)
Because the number of elderly von Willebrand disease (VWD) patients is increasing, the pathophysiology of aging in VWD has become increasingly relevant. To assess age-related changes in von Willebrand factor (VWF) and factor VIII (FVIII) levels and to compare age-related
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service