Skip to Content
Merck
All Photos(1)

Key Documents

RAB0629

Sigma-Aldrich

Human GDF2 / Growth/Differentiation Factor 2 ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$1,150.00

$1,150.00


Check Cart for Availability


Select a Size

Change View
96 WELLS
$1,150.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$1,150.00


Check Cart for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 32 pg/mL
standard curve range: 32.77-8000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GDF2(2658)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
RAB0204RAB0601RAB1481
Gene Information

human ... GDF2(2658)

Gene Information

human ... GDF15(9518)

Gene Information

human ... FGF1(2246)

Gene Information

human ... Gdf3(14562)

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma
sample type urine

input

sample type urine
sample type cell culture supernatant(s)
sample type plasma
sample type serum

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 32 pg/mL
standard curve range: 32.77-8000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 1.1-800 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 12 pg/mL
standard curve range: 16.38-4000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.7 ng/mL
standard curve range: 1.638-400 ng/mL

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

General description

The growth differentiation factor 2 (GDF2) gene encodes for bone morphogenic protein 9 (BMP9).[1] GDF2, also known as BMP9, belongs to the BMP family.[2] The GDF2 gene is mapped on the human chromosome at 10q11.22.[3] GDF2/BMP9 protein regulates angiogenesis, modulating tumorigenesis, inhibiting hepatic glucose production, and maintaining basal forebrain cholinergic neurons.[2] It is implicated in the pathogenesis of pulmonary arterial hypertension.[2] Mutations in the GDF2 gene are associated with hereditary hemorrhagic telangiectasia (HHT), an autosomal dominant vascular disorder.[1] The antibody pair provided in this kit recognizes human growth/differentiation factor 2.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)

  • RABSTOP3ELISA Stop Solution (Item I)

  • RABWASH420X Wash Buffer (Item B)

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Liu et al.
Genes & diseases, 7(2), 235-244 (2020-03-28)
Bone morphogenetic protein 9 (BMP9) (or GDF2) was originally identified from fetal mouse liver cDNA libraries. Emerging evidence indicates BMP9 exerts diverse and pleiotropic functions during postnatal development and in maintaining tissue homeostasis. However, the expression landscape of BMP9 signaling
Felicia Hernandez et al.
Human genome variation, 2, 15040-15040 (2015-01-01)
Hereditary hemorrhagic telangiectasia (HHT) is an autosomal dominant vascular disorder caused by mutations in ENG, ACVRL1 and SMAD4, which function in regulating the transforming growth factor beta and bone morphogenetic protein signaling pathways. Symptoms of HHT can be present in
Z Yang et al.
Science advances, 6(48) (2020-11-29)
Obesity drives the development of nonalcoholic fatty liver disease (NAFLD) characterized by hepatic steatosis. Several bone morphogenetic proteins (BMPs) except BMP9 were reported related to metabolic syndrome. This study demonstrates that liver cytokine BMP9 is decreased in the liver and
Comprehensive copy number variant (CNV) analysis of neuronal pathways genes in psychiatric disorders identifies rare variants within patients.
Saus, et al.
Journal of Psychiatric Research, 44, 971-978 (2018)
Yan-Man Zhou et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 35(5), 978-993 (2020-01-09)
Bone remodeling is dynamic and is tightly regulated through bone resorption dominated by osteoclasts and bone formation dominated by osteoblasts. Imbalances in this process can cause various pathological conditions, such as osteoporosis. Bone morphogenetic protein 9 (BMP9), a biomolecule produced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service