Skip to Content
Merck
All Photos(1)

Key Documents

RAB0612

Sigma-Aldrich

Human APOC2 / Apolipoprotein C-II ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$1,400.00

$1,400.00


Estimated to ship on28 May 2025



Select a Size

Change View
96 WELLS
$1,400.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$1,400.00


Estimated to ship on28 May 2025


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.5 ng/mL
standard curve range: 1.563-100 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... APOC2(344)

General description

Human APOC2 (apolipoprotein C2) is an 8914 kDa exchangeable apolipoprotein, linked with VLDL (very low density lipoprotein) and chylomicrons.[1] This gene is located on human chromosome 19q13.[2]
The ApoC2 ELISA kit provides for the Quantitative measurement of ApoC2 in Cell Culture Supernatants, Plasma and Serum.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

APOC2 (apolipoprotein C2) serves as a cofactor for lipoprotein lipase.[1] It plays a major role in the flux of TGs (triglycerides) through the plasma compartment.[3]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modulation of lipoprotein lipase activity by apolipoproteins. Effect of apolipoprotein C-III.
Wang CS, et al.
The Journal of Clinical Investigation, 75(2), 384-390 (1985)
Alzheimer's disease gene signature says: beware of brain viral infections.
Porcellini E, et al.
Immunity & ageing : I & A, 7(1), 16-16 (2010)
NBD-labeled phospholipid accelerates apolipoprotein C-II amyloid fibril formation but is not incorporated into mature fibrils.
Ryan TM, et al.
Biochemistry, 50(44), 9579-9586 (2011)

Questions

  1. can you send me a copy of the method protocol/kit insert - APOc2 Elisa- RAB0612?

    1 answer
    1. The assay instructions can be found in the kit Certificate of Analysis at the link below. The protocol is not lot specific so the sample Certificate for the kit can be applied to any kit lot:

      https://www.sigmaaldrich.com/product/sigma/rab0612#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service