Skip to Content
Merck
All Photos(1)

Key Documents

RAB0606

Sigma-Aldrich

Human ANGPTL3 / Angiopoietin-related Protein 3 ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$1,020.00

$1,020.00


Estimated to ship on16 April 2025



Select a Size

Change View
96 WELLS
$1,020.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$1,020.00


Estimated to ship on16 April 2025


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8.2 pg/mL
standard curve range: 8.19-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ANGPTL3(27329)

General description

Angiopoietin-related protein 3 (ANGPTL3) is a member of the ANGPTL family and is expressed in the liver.[1] The ANGPTL3 gene is mapped on the human chromosome at 1p31.3.[2] Mutations in the ANGPTL3 gene lead to familial combined hypolipidemia, a Mendelian condition that is indicated by lower levels of triglycerides, high-density lipoprotein (HDL), and low-density lipoprotein cholesterol.[3] The ANGPTL3 enzyme-linked immunosorbent assay (ELISA) kit provides for the quantitative measurement of angiopoietin-related protein 3 in cell culture supernatants, plasma, and serum.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junpeng Ma et al.
Frontiers in oncology, 10, 1382-1382 (2020-09-15)
Meningiomas, as the most common primary tumor of the central nervous system, are known to harbor genomic aberrations that associate with clinical phenotypes. Here we performed genome-wide genotyping for cranial meningiomas in 383 Chinese patients and identified 9,821 copy-number variations
Maria Fernanda Garcés et al.
Scientific reports, 10(1), 21118-21118 (2020-12-05)
This study aimed to determine ANGPTL3 serum levels in healthy young lean and obese non-diabetic men during an oral glucose tolerance test (OGTT) and correlate them with anthropometric, biochemical and hormonal parameters. A case-control study was carried out and 30
Fabiana Quagliarini et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(48), 19751-19756 (2012-11-15)
Angiopoietin-like proteins (ANGPTLs) play major roles in the trafficking and metabolism of lipids. Inactivation of ANGPTL3, a gene located in an intron of DOCK7, results in very low levels of LDL-cholesterol (C), HDL-C and triglyceride (TAG). We identified another ANGPTL
Oleg Kovrov et al.
Journal of lipid research, 60(4), 783-793 (2019-01-29)
Angiopoietin-like (ANGPTL) 8 is a secreted inhibitor of LPL, a key enzyme in plasma triglyceride metabolism. It was previously reported that ANGPTL8 requires another member of the ANGPTL family, ANGPTL3, to act on LPL. ANGPTL3, much like ANGPTL4, is a
Andrew A Butler et al.
Journal of lipid research, 61(3), 376-386 (2020-01-11)
Angiopoietin-like protein 3 (ANGPTL3) inhibits lipid clearance and is a promising target for managing cardiovascular disease. Here we investigated the effects of a high-sugar (high-fructose) diet on circulating ANGPTL3 concentrations in rhesus macaques. Plasma ANGPTL3 concentrations increased ∼30% to 40%

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service