Skip to Content
Merck
All Photos(1)

Key Documents

RAB0579

Sigma-Aldrich

Canine TIMP2 / Metalloproteinase Inhibitor 2 ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$970.00

$970.00


Estimated to ship on01 June 2025



Select a Size

Change View
96 WELLS
$970.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$970.00


Estimated to ship on01 June 2025


species reactivity

canine

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 20.48-5000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

canine ... TIMP2(403633)

General description

The antibody pair provided in this kit recognizes Canine Metalloproteinase inhibitor 2.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service