Skip to Content
Merck
All Photos(1)

Documents

RAB0500

Sigma-Aldrich

Human p53 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type urine
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.3 ng/mL
standard curve range: 0.328-80 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TP53(7157)

General description

Cellular tumor antigen p53 is a tumor-suppressor protein that comprises transactivation at N-terminus, central sequence-specific DNA-binding domain, and regulatory domain at C-terminus. It plays a key role in autophagy regulation. It inhibits tumors and acts as a cellular stress sensor especially during nutrient deprivation, hypoxia, and ribosome dysfunction. The Human p-53 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human p-53 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human p53

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Human p53 ELISA Kit has been used for quantifying p53 using enzyme-linked immunosorbent assay (ELISA) in human myeloma RPMI-8226 cells, human colorectal cancer HCT-116 cells, human breast cancer MCF-7 cells and human leukemiaHL-60 cells.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADDELISA 5X Assay/Sample Diluent Buffer D (Item K)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of roscovitine anticancer agent as a reference compound for cancer and apoptosis studies
Taha H, et al.
JIMD Reports null
Mohammed K Abd Elhameid et al.
Chemical & pharmaceutical bulletin, 66(10), 939-952 (2018-08-17)
A new series of pyridine and pyrimidine derivatives is designed and synthesized as potential antitumor molecules. The tested compounds show promising in vitro cytotoxic activity against HL-60 cell line as eight compounds: 4, 6, 11, 13, 14, 15, 18 and
p53 at a glance.
Colleen A Brady et al.
Journal of cell science, 123(Pt 15), 2527-2532 (2010-10-14)
Wei Li et al.
Oncology letters, 9(4), 1719-1724 (2015-03-20)
Curcumin is an inexpensive, natural plant ingredient with protease inhibitor effects. The present study aimed to analyze the inhibitory effects of curcumin on the multiple myeloma (MM) RPMI 8226 cell line, and examine the underlying mechanism that promotes the apoptosis
Walaa Hamada Abd-Allah et al.
Drug development research, 80(7), 933-947 (2019-07-26)
Novel 1,1-disubstituted cyclohexane-1-carboxamides 6a-h, 7a-e, and 8a-b were designed and synthesized as apoptotic inducers. Cytotoxicity test revealed that some compounds have strong to moderate effect, while others displayed weak action against different cancer cell lines including, MCF-7, HepG2, A549, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service