Skip to Content
Merck
All Photos(2)

Key Documents

RAB0494

Sigma-Aldrich

Mouse TRANCE ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$915.00

$915.00


Check Cart for Availability


Select a Size

Change View
1 KIT
$915.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$915.00


Check Cart for Availability

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

The Mouse Trance ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse Trance in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse TRANCE

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Huang et al.
Bone, 66, 306-314 (2014-06-17)
Reactive oxygen species (ROS) are a significant pathogenic factor of osteoporosis. Ginsenoside-Rb2 (Rb2), a 20(S)-protopanaxadiol glycoside extracted from ginseng, is a potent antioxidant that generates interest regarding the bone metabolism area. We tested the potential anti-osteoporosis effects of Rb2 and
Jiang Lin et al.
Infection and immunity, 82(10), 4127-4134 (2014-07-23)
Toll-like receptors (TLRs) play a key role in the innate immune responses to periodontal pathogens in periodontal disease. The present study was performed to determine the roles of TLR2 and TLR4 signaling in alveolar bone resorption, using a Porphyromonas gingivalis-associated
Regulation of Osteoblast Differentiation by Acid-Etched and/or Grit-Blasted Titanium Substrate Topography Is Enhanced by 1,25(OH)2D3 in a Sex-Dependent Manner.
Olivares-Navarrete R, Hyzy SL, Boyan BD, et al.
BioMed Research International, 2015, 365014-365014 (2015)
Stella Bernardi et al.
Molecular and cellular endocrinology, 394(1-2), 13-20 (2014-07-08)
Inflammation is believed to link obesity to insulin resistance, as in the setting of metabolic syndrome (MetS). Osteoprotegerin (OPG) is a soluble protein that seems to exert proatherogenic and prodiabetogenic effects. This study aims at determining OPG levels in MetS
C A Simpson et al.
Osteoporosis international : a journal established as result of cooperation between the European Foundation for Osteoporosis and the National Osteoporosis Foundation of the USA, 25(10), 2383-2388 (2014-06-15)
We compared circulating levels of Wnt inhibitors among patients with high bone mass mutations in LRP5, unaffected kindred, and unrelated normal controls. Inhibitors were unchanged in affected and unaffected kindred. We saw no meaningful differences between controls and affected individuals.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service