Skip to Content
Merck
All Photos(2)

Documents

RAB0388

Sigma-Aldrich

Human NRG1-β 1 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type urine
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 50 pg/mL
standard curve range: 82.3-20000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NRG1(3084)

General description

The Human NRG1-β 1 (Neuregulin 1-β 1) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human NRG1-β 1 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human NRG-1 β-1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi Du et al.
Journal of proteome research, 18(5), 2021-2031 (2019-03-26)
For individuals migrating to or residing permanently in high-altitude regions, environmental hypobaric hypoxia is a primary challenge that induces several physiological or pathological responses. It is well documented that human beings adapt to hypobaric hypoxia via some protective mechanisms, such
Charline Ogier et al.
Cancer letters, 432, 227-236 (2018-06-24)
Neuregulin 1 (NRG1), a ligand for HER3 and HER4 receptors, is secreted by both pancreatic tumor cells (PC) and cancer-associated fibroblasts (CAFs), the latter representing the most abundant compound of pancreatic stroma. This desmoplastic stroma contributes to Pancreatic Ductal Adenocarcinoma
Camilla Hage et al.
ESC heart failure, 7(2), 445-455 (2020-01-26)
Neuregulin1-β (NRG1-β) is released from microvascular endothelial cells in response to inflammation with compensatory cardioprotective effects. Circulating NRG1-β is elevated in heart failure (HF) with reduced ejection fraction (HFrEF) but not studied in HF with preserved EF (HFpEF). Circulating NRG1-β
Kimio Yonesaka et al.
Scientific reports, 9(1), 19501-19501 (2019-12-22)
Epidermal growth factor receptor-tyrosine kinase inhibitors (EGFR-TKIs) are standard therapy for EGFR-mutant non-small cell lung cancer (NSCLC). Preclinically, HER3 ligand heregulin induces resistance to EGFR-TKIs, whereas the pan-human EGFR family inhibitor afatinib remains effective. Here, we examined whether soluble heregulin
Giulia Ronchi et al.
BioMed research international, 2018, 9175248-9175248 (2018-07-17)
Muscle-in-vein conduit is successfully employed for repairing nerve injuries: the vein prevents muscle fiber dispersion, while the muscle prevents the vein collapse and creates a favorable environment for Schwann cell migration and axon regrowth. However, it requires microsurgical skills. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service