Skip to Content
Merck
All Photos(2)

Key Documents

RAB0303

Sigma-Aldrich

Human IL-5 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$918.00

$918.00


Estimated to ship on01 June 2025



Select a Size

Change View
1 KIT
$918.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$918.00


Estimated to ship on01 June 2025


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type urine
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL5(3567)

General description

Interleukin 1 (IL-5) exists as a homodimer and elicits its cytokine functions via interleukin receptor (IL-5R). It mediates the maturation of eosinophils. IL-5 also exhibits pleiotropic effects on B cells favoring cell survival and differentiation. Due to its key role in eosinophilia and immune response, IL-5 serves as a potential therapeutic target in treating eosinophil-mediated disorders.[1] The Human IL-5 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human interleukin (IL-5) in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human IL-5

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taku Kouro et al.
International immunology, 21(12), 1303-1309 (2009-10-13)
IL-5 was originally defined as a T-cell-derived cytokine that triggers activated B cells for terminal differentiation into antibody-secreting plasma cells, at least in mice. Concurrently, IL-5 was recognized as the major maturation and differentiation factor for eosinophils in mice and
P N Adams et al.
Parasite immunology, 36(10), 531-539 (2014-07-22)
The M2 subset of macrophages has a critical role to play in host tissue repair, tissue fibrosis and modulation of adaptive immunity during helminth infection. Infection with the helminth, Fasciola hepatica, is associated with M2 macrophages in its mammalian host
Ershun Zhou et al.
Fitoterapia, 96, 131-137 (2014-05-03)
Thymol, a naturally occurring monocyclic phenolic compound derived from Thymus vulgaris (Lamiaceae), has been reported to exhibit anti-inflammatory property in vivo and vitro. However, the mechanism of thymol is not clear. The aim of the present study was to investigate
Sean O Ryan et al.
Journal of immunology (Baltimore, Md. : 1950), 192(12), 5561-5570 (2014-05-06)
Common variable immunodeficiency (CVID), the most frequent symptomatic primary immune deficiency in humans, is a heterogeneous group of immunologic disorders estimated to affect 1:10,000-1:50,000. Although a clear disease etiology remains elusive, a common characteristic of CVID is deficient IgG Ab

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service