Skip to Content
Merck
All Photos(2)

Key Documents

RAB0289

Sigma-Aldrich

Mouse IL-21 ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$915.00

$915.00


Check Cart for Availability


Select a Size

Change View
1 KIT
$915.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$915.00


Check Cart for Availability

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 8.19-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il21(60505)

General description

The Mouse IL-21 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse IL-21 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse IL-21

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng Cheng et al.
International journal of clinical and experimental medicine, 7(10), 3202-3208 (2014-11-25)
Asthma is characterized by airway inflammation, mucus overproduction, and airway hyperreactivity. Cytokines, especially T helper 2-derived cytokines interleukin (IL)-4, IL-5, and IL-13, are involved in the pathogenesis of asthma. IL-21 has a variety of effects on the immune system. However
Mohan S Maddur et al.
Nature communications, 5, 4092-4092 (2014-06-10)
Dendritic cells (DCs) play a critical role in immune homeostasis by regulating the functions of various immune cells, including T and B cells. Notably, DCs also undergo education on reciprocal signalling by these immune cells and environmental factors. Various reports
Li Wang et al.
The Journal of rheumatology, 41(9), 1781-1792 (2014-07-17)
To elucidate the potential role of follicular helper T cells (TFH) and interleukin 10 (IL-10)+ B cells in the development of systemic lupus erythematosus (SLE). The numbers of peripheral blood CD27+, CD38+, CD86+, CD95+, IL-10+ B cells, and inducible T

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service